COM448631566 Combi-Blocks, Inc.

N-Benzyl L-isoleucinamide

  • MDL number MFCD14590299
  • Cas Number : 129253-02-7
  • Molecular Weight : 220.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2464
Availability details for
Quantity
  • Availability
  • Add to Cart