COM448631585 Combi-Blocks, Inc.

N-(2-Aminophenyl)-N-methylmethanesulfonamide

  • MDL number MFCD16158909
  • Cas Number : 90140-21-9
  • Molecular Weight : 200.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2489

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2489
Availability details for
Quantity
  • Availability
  • Add to Cart