COM448631589 Combi-Blocks, Inc.

4-Imino-5-methylheptan-2-one

  • MDL number MFCD25953913
  • Cas Number : 1823377-64-5
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2494
Availability details for
Quantity
  • Availability
  • Add to Cart