COM448631663 Combi-Blocks, Inc.

Methyl 4-(methanesulfonylmethyl)benzoate

  • MDL number MFCD00051693
  • Cas Number : 160446-22-0
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2633
Availability details for
Quantity
  • Availability
  • Add to Cart