COM448631667 Combi-Blocks, Inc.

3-Butoxy-4-fluorobenzaldehyde

  • MDL number MFCD18426723
  • Cas Number : 1378815-44-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2638
Availability details for
Quantity
  • Availability
  • Add to Cart