COM448631668 Combi-Blocks, Inc.

3-(Cyclopropylmethoxy)-4-fluorobenzaldehyde

  • MDL number MFCD24675206
  • Cas Number : 1208231-17-7
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2640

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2640
Availability details for
Quantity
  • Availability
  • Add to Cart