COM448631669 Combi-Blocks, Inc.

3-Cyclobutoxy-4-fluorobenzaldehyde

  • MDL number MFCD28101640
  • Cas Number : 1820606-99-2
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2643

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2643
Availability details for
Quantity
  • Availability
  • Add to Cart