COM448631670 Combi-Blocks, Inc.

3-(Cyclohexyloxy)-4-fluorobenzaldehyde

  • MDL number MFCD28101641
  • Cas Number : 1289156-60-0
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2644

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2644
Availability details for
Quantity
  • Availability
  • Add to Cart