COM448631710 Combi-Blocks, Inc.

3-Bromo-2-(trifluoromethyl)phenol

  • MDL number MFCD13194489
  • Cas Number : 1214352-26-7
  • Molecular Weight : 241.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2706

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2706
Availability details for
Quantity
  • Availability
  • Add to Cart