COM448631732 Combi-Blocks, Inc.

2-(4-Methylpiperazine-1-sulfonyl)ethanamine hydrochloride

  • MDL number MFCD27935909
  • Cas Number : 1820706-33-9
  • Molecular Weight : 243.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2732
Availability details for
Quantity
  • Availability
  • Add to Cart