COM448631733 Combi-Blocks, Inc.

3,4-Dihydro-1,3-benzoxazin-2-one

  • MDL number MFCD02313739
  • Cas Number : 1125-85-5
  • Molecular Weight : 149.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2733
Availability details for
Quantity
  • Availability
  • Add to Cart