COM448631737 Combi-Blocks, Inc.

1-(4-Bromophenyl)pent-4-en-1-ol

  • MDL number MFCD20290882
  • Cas Number : 346446-95-5
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2738

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2738
Availability details for
Quantity
  • Availability
  • Add to Cart