COM448631745 Combi-Blocks, Inc.

Amino(2-methylphenyl)acetic acid hydrochloride

  • MDL number MFCD06410551
  • Cas Number : 879500-49-9
  • Molecular Weight : 201.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2755
Availability details for
Quantity
  • Availability
  • Add to Cart