COM448631746 Combi-Blocks, Inc.

1-(Benzyloxy)-2-bromo-3-(trifluoromethyl)benzene

  • MDL number MFCD27981317
  • Cas Number : 1820703-43-2
  • Molecular Weight : 331.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2756
Availability details for
Quantity
  • Availability
  • Add to Cart