COM448631753 Combi-Blocks, Inc.

N-(4-Bromophenethyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD03854862
  • Cas Number : 181514-21-6
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2768
Availability details for
Quantity
  • Availability
  • Add to Cart