COM448631758 Combi-Blocks, Inc.

[2-Amino-1-(3-bromophenyl)ethyl]dimethylamine

  • MDL number MFCD09727568
  • Cas Number : 951917-59-2
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2777
Availability details for
Quantity
  • Availability
  • Add to Cart