COM448631771 Combi-Blocks, Inc.

8-Chlorooct-1-yne

  • MDL number MFCD07656663
  • Cas Number : 24088-97-9
  • Molecular Weight : 144.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2802

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2802
Availability details for
Quantity
  • Availability
  • Add to Cart