COM448631773 Combi-Blocks, Inc.

3-(Benzyloxy)-4-bromotoluene

  • MDL number MFCD28014039
  • Cas Number : 177555-85-0
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2805

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2805
Availability details for
Quantity
  • Availability
  • Add to Cart