COM448631777 Combi-Blocks, Inc.

2-(4-Chloro-2-fluorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD28014040
  • Cas Number : 1820711-18-9
  • Molecular Weight : 198.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2812

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2812
Availability details for
Quantity
  • Availability
  • Add to Cart