COM448631778 Combi-Blocks, Inc.

5-(4-Chloro-2-fluorophenyl)-3H-1,3,4-oxadiazol-2-one

  • MDL number MFCD19673382
  • Cas Number : 1342016-50-5
  • Molecular Weight : 214.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2813
Availability details for
Quantity
  • Availability
  • Add to Cart