COM448631781 Combi-Blocks, Inc.

[2-Amino-1-(2-fluorophenyl)ethyl]dimethylamine

  • MDL number MFCD06753110
  • Cas Number : 842954-76-1
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2816

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2816
Availability details for
Quantity
  • Availability
  • Add to Cart