COM448631786 Combi-Blocks, Inc.

5-Fluoro-2-isopropoxybenzonitrile

  • MDL number MFCD15201676
  • Cas Number : 1158506-01-4
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2826
Availability details for
Quantity
  • Availability
  • Add to Cart