COM448631789 Combi-Blocks, Inc.

(R)-(2-Cyano-1-methylethyl)carbamic acid, 1,1-dimethylethyl ester

  • MDL number MFCD12197316
  • Cas Number : 170367-68-7
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2829

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2829
Availability details for
Quantity
  • Availability
  • Add to Cart