COM448631795 Combi-Blocks, Inc.

(2-Fluorophenyl)glycine HCl

  • MDL number MFCD28096639
  • Cas Number : 709665-70-3
  • Molecular Weight : 205.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2836

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2836
Availability details for
Quantity
  • Availability
  • Add to Cart