COM448631798 Combi-Blocks, Inc.

4-(Pyrrolidinocarbonyl)phenol

  • MDL number MFCD07633880
  • Cas Number : 478929-28-1
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2839
Availability details for
Quantity
  • Availability
  • Add to Cart