COM448631802 Combi-Blocks, Inc.

Benzyl 2-fluoro-6-nitrobenzoate

  • MDL number MFCD28096640
  • Cas Number : 1820604-19-0
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2843

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2843
Availability details for
Quantity
  • Availability
  • Add to Cart