COM448631811 Combi-Blocks, Inc.

1-Bromo-3-chloro-5-(chloromethyl)benzene

  • MDL number MFCD13189468
  • Cas Number : 1060812-67-0
  • Molecular Weight : 239.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2855

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2855
Availability details for
Quantity
  • Availability
  • Add to Cart