COM448631816 Combi-Blocks, Inc.

2-(2,3-Dichlorophenoxy)acetohydrazide

  • MDL number MFCD00245649
  • Cas Number : 153860-25-4
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2860

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2860
Availability details for
Quantity
  • Availability
  • Add to Cart