COM448631819 Combi-Blocks, Inc.

Diethyl 2-[(4-bromo-3-methylphenyl)aminomethylidene]malonate

  • MDL number MFCD28101643
  • Cas Number : 1820686-69-8
  • Molecular Weight : 356.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2866
Availability details for
Quantity
  • Availability
  • Add to Cart