COM448631826 Combi-Blocks, Inc.

N-Methyl-N-(4-nitrophenyl)methanesulfonamide

  • MDL number MFCD25962440
  • Cas Number : 334952-01-1
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2873

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2873
Availability details for
Quantity
  • Availability
  • Add to Cart