COM448631835 Combi-Blocks, Inc.

1-(4-Chloro-2-hydroxy-5-methoxyphenyl)thiourea

  • MDL number MFCD28101645
  • Cas Number : 1820683-26-8
  • Molecular Weight : 232.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2882
Availability details for
Quantity
  • Availability
  • Add to Cart