COM448631838 Combi-Blocks, Inc.

(N-BOC-Amino)(naphthalen-1-yl)acetic acid

  • MDL number MFCD07388849
  • Cas Number : 14675-99-1
  • Molecular Weight : 301.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2885

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2885
Availability details for
Quantity
  • Availability
  • Add to Cart