COM448631841 Combi-Blocks, Inc.

2-[Benzyl(2-chloroethyl)amino]ethanol hydrochloride

  • MDL number MFCD28122690
  • Cas Number : 137834-82-3
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2888

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2888
Availability details for
Quantity
  • Availability
  • Add to Cart