COM448631845 Combi-Blocks, Inc.

(4-Bromo-2-hydroxyphenyl)thiourea

  • MDL number MFCD22483179
  • Cas Number : 1404480-07-4
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2894

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2894
Availability details for
Quantity
  • Availability
  • Add to Cart