COM448631847 Combi-Blocks, Inc.

3-(2-Hydroxy-2-phenylethyl)-1,3-oxazolidin-2-one

  • MDL number MFCD16042772
  • Cas Number : 82191-11-5
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2896

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2896
Availability details for
Quantity
  • Availability
  • Add to Cart