COM448631851 Combi-Blocks, Inc.

N-(4-Aminophenyl)-N-methylmethanesulfonamide

  • MDL number MFCD11184922
  • Cas Number : 251552-20-2
  • Molecular Weight : 200.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2904

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2904
Availability details for
Quantity
  • Availability
  • Add to Cart