COM448631852 Combi-Blocks, Inc.

N-Butyl-3-[(hydrazinecarbonyl)methoxy]benzamide

  • MDL number MFCD28369587
  • Cas Number : 1820735-59-8
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2905
Availability details for
Quantity
  • Availability
  • Add to Cart