COM448631854 Combi-Blocks, Inc.

2-(4-Fluoro-2-nitrophenoxy)acetohydrazide

  • MDL number MFCD18369703
  • Cas Number : 1341119-52-5
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2907

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2907
Availability details for
Quantity
  • Availability
  • Add to Cart