COM448631859 Combi-Blocks, Inc.

3-(1,3-Dioxoisoindol-2-yl)benzaldehyde

  • MDL number MFCD00512270
  • Cas Number : 205759-45-1
  • Molecular Weight : 251.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2913

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2913
Availability details for
Quantity
  • Availability
  • Add to Cart