COM448631862 Combi-Blocks, Inc.

2-Bromo-4-ethoxy-1-fluorobenzene

  • MDL number MFCD25318992
  • Cas Number : 1555379-66-2
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2919
Availability details for
Quantity
  • Availability
  • Add to Cart