COM448631868 Combi-Blocks, Inc.

N-Methyl-2,3-dihydro-1,4-benzodioxine-6-carboxamide

  • MDL number MFCD02713088
  • Cas Number : 438026-68-7
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2933
Availability details for
Quantity
  • Availability
  • Add to Cart