COM448631871 Combi-Blocks, Inc.

1-{[(tert-Butoxy)carbonyl]amino}-2-ethenylcyclopropane-1-carboxylic acid

  • MDL number MFCD14525711
  • Cas Number : 1001667-24-8
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2936

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2936
Availability details for
Quantity
  • Availability
  • Add to Cart