COM448631879 Combi-Blocks, Inc.

3-Iodo-L-tyrosine methyl ester hydrochloride

  • MDL number MFCD07778603
  • Cas Number : 79677-58-0
  • Molecular Weight : 357.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2949

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2949
Availability details for
Quantity
  • Availability
  • Add to Cart