COM448631886 Combi-Blocks, Inc.

Ethyl 7-bromo-2-oxochromene-3-carboxylate

  • MDL number MFCD22373079
  • Cas Number : 105837-04-5
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2964

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2964
Availability details for
Quantity
  • Availability
  • Add to Cart