COM448631888 Combi-Blocks, Inc.

4-Bromo-2-ethoxy-1-iodobenzene

  • MDL number MFCD12547814
  • Cas Number : 943742-29-8
  • Molecular Weight : 327.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2967
Availability details for
Quantity
  • Availability
  • Add to Cart