COM448631889 Combi-Blocks, Inc.

tert-Butyl N-(2,3-dihydro-1H-inden-2-yl)carbamate

  • MDL number MFCD24465808
  • Cas Number : 1190890-29-9
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2974

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2974
Availability details for
Quantity
  • Availability
  • Add to Cart