COM448631895 Combi-Blocks, Inc.

1-(4-Bromophenyl)prop-2-en-1-one

  • MDL number MFCD02259860
  • Cas Number : 22731-70-0
  • Molecular Weight : 211.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2989

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2989
Availability details for
Quantity
  • Availability
  • Add to Cart