COM448631898 Combi-Blocks, Inc.

D-Fmoc-Ornithine HCl

  • MDL number MFCD28252211
  • Cas Number : 1820569-07-0
  • Molecular Weight : 390.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2994

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2994
Availability details for
Quantity
  • Availability
  • Add to Cart