COM448631900 Combi-Blocks, Inc.

5-Bromo-2-ethenylphenol

  • MDL number MFCD28252212
  • Cas Number : 1510865-23-2
  • Molecular Weight : 199.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2996

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2996
Availability details for
Quantity
  • Availability
  • Add to Cart