COM448631902 Combi-Blocks, Inc.

3-(Trifluoromethoxy)-dl-phenylglycine HCl

  • MDL number MFCD28369588
  • Cas Number : 1820603-68-6
  • Molecular Weight : 271.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2998
Availability details for
Quantity
  • Availability
  • Add to Cart